Anti-CAPRIN1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CAPRIN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-512
- 77822-513
- Antigen symbol:
- CAPRIN1
- CAPRIN1
- Antigen name:
- Caprin-1 protein (573-703AA)
- Caprin-1 protein (573-703AA)
- Antigen synonyms:
- activation/proliferation-associated protein 1|CAPR1_HUMAN|Caprin-1|CAPRIN1|Cell cycle-associated protein 1|Cytoplasmic activation- and proliferation-associated protein 1|cytoplasmic activation/proliferation-associated protein-1|GPI-anchored membrane protein 1|GPI-anchored protein p137|GPI-p137|GPIAP1|GPIP137|M11S1|Membrane component chromosome 11 surface marker 1|p137GPI|RNA granule protein 105|RNG105
- activation/proliferation-associated protein 1|CAPR1_HUMAN|Caprin-1|CAPRIN1|Cell cycle-associated protein 1|Cytoplasmic activation- and proliferation-associated protein 1|cytoplasmic activation/proliferation-associated protein-1|GPI-anchored membrane protein 1|GPI-anchored protein p137|GPI-p137|GPIAP1|GPIP137|M11S1|Membrane component chromosome 11 surface marker 1|p137GPI|RNA granule protein 105|RNG105
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14444
- Q14444
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together