Anti-PCOTH Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCOTH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-862
- 77826-861
- Antigen symbol:
- PCOTH
- PCOTH
- Antigen name:
- Prostate collagen triple helix protein (50-100AA)
- Prostate collagen triple helix protein (50-100AA)
- Antigen synonyms:
- C1QTNF9B antisense RNA 1 (non-protein coding)|PCOTH_HUMAN|Prostate collagen triple helix protein|Protein PCOTH
- C1QTNF9B antisense RNA 1 (non-protein coding)|PCOTH_HUMAN|Prostate collagen triple helix protein|Protein PCOTH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q58A44
- Q58A44
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together