Anti-JAK2 Rabbit Polyclonal Antibody
Catalog # 77813-082
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Tyrosine-protein kinase JAK2
- Antigen symbol:JAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Size:100 µl
- Form:liquid
- Gene ID:O60674
- Antigen synonyms:JAK 2|JAK-2|JAK2|JAK2_HUMAN|Janus Activating Kinase 2|Janus kinase 2 (a protein tyrosine kinase)|Janus kinase 2|JTK 10|JTK10|kinase Jak2|OTTHUMP00000043260|THCYT3|Tyrosine protein kinase JAK2|Tyrosine-protein kinase JAK2
- Storage buffer:PBS, 150mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:100
Type:
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Rat