Anti-HLA-C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HLA-C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77815-222
- 77815-221
- Antigen symbol:
- HLA-C
- HLA-C
- Antigen name:
- HLA class I histocompatibility antigen, Cw-7 alpha chain protein (62-306AA)
- HLA class I histocompatibility antigen, Cw-7 alpha chain protein (62-306AA)
- Antigen synonyms:
- 1C07_HUMAN|Cw-7 alpha chain|D6S204|HLA class I histocompatibility antigen|C alpha chain|Cw-1 alpha chain|HLA JY3|HLA-C|HLA-C histocompatibility type|HLA-Cw|HLA-JY3|HLAC|HLC C|HLC-C|human leukocyte antigen-C alpha chain|major histocompatibility antigen HLA-C|major histocompatibility complex|class I|C |MHC class I antigen|MHC class I antigen Cw 1|MHC class I antigen Cw*7|MHC class I antigen heavy chain HLA-C|PSORS1
- 1C07_HUMAN|Cw-7 alpha chain|D6S204|HLA class I histocompatibility antigen|C alpha chain|Cw-1 alpha chain|HLA JY3|HLA-C|HLA-C histocompatibility type|HLA-Cw|HLA-JY3|HLAC|HLC C|HLC-C|human leukocyte antigen-C alpha chain|major histocompatibility antigen HLA-C|major histocompatibility complex|class I|C |MHC class I antigen|MHC class I antigen Cw 1|MHC class I antigen Cw*7|MHC class I antigen heavy chain HLA-C|PSORS1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P10321
- P10321
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together