Anti-wnt8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: wnt8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Xenopus laevis
Specifications
- Catalog No:
- 77816-336
- 77816-337
- Antigen symbol:
- wnt8
- wnt8
- Antigen name:
- Recombinant Xenopus laevis Protein Wnt-8 protein (23-358AA)
- Recombinant Xenopus laevis Protein Wnt-8 protein (23-358AA)
- Antigen synonyms:
- wnt8|Protein Wnt-8|XWnt-8
- wnt8|Protein Wnt-8|XWnt-8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Xenopus laevis
- Xenopus laevis
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28026
- P28026
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together