Anti-KCNK13 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNK13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-702
- 77835-701
- Antigen symbol:
- KCNK13
- KCNK13
- Antigen name:
- Potassium channel subfamily K member 13 protein (292-408AA)
- Potassium channel subfamily K member 13 protein (292-408AA)
- Antigen synonyms:
- K2p13.1|K2P13.1 potassium channel|Kcnk13|KCNKD|KCNKD_HUMAN|Potassium channel subfamily K member 13|Tandem pore domain halothane inhibited potassium channel 1|Tandem pore domain halothane-inhibited potassium channel 1|Tandem pore domain potassium channel THIK 1|Tandem pore domain potassium channel THIK1|Thik 1|THIK-1|Thik1
- K2p13.1|K2P13.1 potassium channel|Kcnk13|KCNKD|KCNKD_HUMAN|Potassium channel subfamily K member 13|Tandem pore domain halothane inhibited potassium channel 1|Tandem pore domain halothane-inhibited potassium channel 1|Tandem pore domain potassium channel THIK 1|Tandem pore domain potassium channel THIK1|Thik 1|THIK-1|Thik1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HB14
- Q9HB14
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together