Anti-SLC14A1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC14A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-881
- 77822-882
- Antigen symbol:
- SLC14A1
- SLC14A1
- Antigen name:
- Urea transporter 1 protein (1-51AA)
- Urea transporter 1 protein (1-51AA)
- Antigen synonyms:
- blood group Kidd urea transporter|erythrocyte|HsT1341|HUT11|JK|Kidd|kidd (JK) blood group urea transporter B1|RACH1|Slc14a1|solute carrier family 14 (urea transporter) member 1 (Kidd blood group)|Solute carrier family 14 member 1|Urea transporter 1|Urea transporter|urea transporter B1|Urea transporter erythrocyte|urea transporter JK glycoprotein|UT B1|UT1|UT1_HUMAN|UTE
- blood group Kidd urea transporter|erythrocyte|HsT1341|HUT11|JK|Kidd|kidd (JK) blood group urea transporter B1|RACH1|Slc14a1|solute carrier family 14 (urea transporter) member 1 (Kidd blood group)|Solute carrier family 14 member 1|Urea transporter 1|Urea transporter|urea transporter B1|Urea transporter erythrocyte|urea transporter JK glycoprotein|UT B1|UT1|UT1_HUMAN|UTE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13336
- Q13336
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together