Anti-AMA-1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AMA-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Specifications
- Catalog No:
- 77816-191
- 77816-190
- Antigen symbol:
- AMA-1
- AMA-1
- Antigen name:
- Recombinant Plasmodium falciparum Apical membrane antigen 1 protein (25-546AA)
- Recombinant Plasmodium falciparum Apical membrane antigen 1 protein (25-546AA)
- Antigen synonyms:
- AMA-1|PF83Apical membrane antigen 1|Merozoite surface antigen
- AMA-1|PF83Apical membrane antigen 1|Merozoite surface antigen
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Plasmodium falciparum
- Plasmodium falciparum
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22621
- P22621
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together