Anti-TXK Rabbit Polyclonal Antibody
77811-737
New Product
- Antigen name:Tyrosine-protein kinase TXK
- Antigen symbol:TXK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:P42681
- Antigen synonyms:BTKL| EC 2.7.10.2| MGC22473| Protein-tyrosine kinase 4| PSCTK5| PTK4| Resting lymphocyte kinase| RLK| TKL| TXK| TXK tyrosine kinase| TXK_HUMAN| Tyrosine protein kinase TXK| Tyrosine-protein kinase TXK
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TXK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human