Anti-GPR63 Rabbit Polyclonal Antibody
77809-315
New Product
- Antigen name:Synthesized peptide derived from the C-terminal region of GPR63
- Antigen symbol:GPR63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BZJ6
- Antigen synonyms:Brain expressed G-protein-coupled receptor PSP24 beta|G protein-coupled receptor 63|G Protein-Coupled Receptor GPR63|Gpr63|GPR63_HUMAN|Probable G-protein coupled receptor 63|PSP24(beta)|PSP24-2|PSP24-beta|PSP24B
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: GPR63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse