Anti-ACOT6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACOT6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-322
- 77826-323
- Antigen symbol:
- ACOT6
- ACOT6
- Antigen name:
- Putative acyl-coenzyme A thioesterase 6 protein (1-207AA)
- Putative acyl-coenzyme A thioesterase 6 protein (1-207AA)
- Antigen synonyms:
- ACOT6| C14orf42Putative acyl-coenzyme A thioesterase 6| Putative acyl-CoA thioesterase 6| EC 3.1.2.-
- ACOT6| C14orf42Putative acyl-coenzyme A thioesterase 6| Putative acyl-CoA thioesterase 6| EC 3.1.2.-
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3I5F7
- Q3I5F7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together