Anti-ldcA Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ldcA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Specifications
- Catalog No:
- 77814-617
- 77814-616
- Antigen symbol:
- ldcA
- ldcA
- Antigen name:
- Recombinant Escherichia coli Murein tetrapeptide carboxypeptidase protein (1-304AA)
- Recombinant Escherichia coli Murein tetrapeptide carboxypeptidase protein (1-304AA)
- Antigen synonyms:
- ldcA|ycgQ|b1192|JW1181Murein tetrapeptide carboxypeptidase|EC 3.4.17.13|LD-carboxypeptidase A|Muramoyltetrapeptide carboxypeptidase
- ldcA|ycgQ|b1192|JW1181Murein tetrapeptide carboxypeptidase|EC 3.4.17.13|LD-carboxypeptidase A|Muramoyltetrapeptide carboxypeptidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P76008
- P76008
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together