Anti-HECTD2 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HECTD2
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-994
- 77828-995
- Antigen symbol:
- HECTD2
- HECTD2
- Antigen name:
- Probable E3 ubiquitin-protein ligase HECTD2 protein (674-776AA)
- Probable E3 ubiquitin-protein ligase HECTD2 protein (674-776AA)
- Antigen synonyms:
- FLJ16050|FLJ37306|HECD2_HUMAN|HECT domain containing 2|HECT domain containing protein 2|HECT domain-containing protein 2|Hectd2|OTTHUMP00000020086|OTTHUMP00000038605|Probable E3 ubiquitin protein ligase HECTD2|Probable E3 ubiquitin-protein ligase HECTD2
- FLJ16050|FLJ37306|HECD2_HUMAN|HECT domain containing 2|HECT domain containing protein 2|HECT domain-containing protein 2|Hectd2|OTTHUMP00000020086|OTTHUMP00000038605|Probable E3 ubiquitin protein ligase HECTD2|Probable E3 ubiquitin-protein ligase HECTD2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5U5R9
- Q5U5R9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together