Anti-VPS25 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VPS25
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-820
- 77842-821
- Antigen symbol:
- VPS25
- VPS25
- Antigen name:
- Vacuolar protein-sorting-associated protein 25 protein (1-176AA)
- Vacuolar protein-sorting-associated protein 25 protein (1-176AA)
- Antigen synonyms:
- Dermal papilla derived protein 9|Dermal papilla-derived protein 9|DERP9|EAP20|ELL associated protein of 20 kDa|ELL-associated protein of 20 kDa|ESCRT-II complex subunit FAP20|HGNC:28122|hMGC10540|Vacuolar protein sorting 25 homolog (S. cerevisiae)|Vacuolar protein sorting associated protein 25|Vacuolar protein-sorting-associated protein 25|VPS25_HUMAN
- Dermal papilla derived protein 9|Dermal papilla-derived protein 9|DERP9|EAP20|ELL associated protein of 20 kDa|ELL-associated protein of 20 kDa|ESCRT-II complex subunit FAP20|HGNC:28122|hMGC10540|Vacuolar protein sorting 25 homolog (S. cerevisiae)|Vacuolar protein sorting associated protein 25|Vacuolar protein-sorting-associated protein 25|VPS25_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRG1
- Q9BRG1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together