Order Entry
Canada
ContactUsLinkComponent
Anti-SOX8 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-SOX8 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SOX8 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77800-757
  • 77800-758
  • Antigen symbol:
  • SOX8
  • SOX8
  • Antigen name:
  • Transcription factor SOX-8 protein (292-386AA)
  • Transcription factor SOX-8 protein (292-386AA)
  • Antigen synonyms:
  • MGC24837|SOX 8|Sox8|SOX8_HUMAN|SRY (sex determining region Y) box 8|SRY box 8|SRY box containing gene 8|Transcription factor SOX 8|Transcription factor SOX-8|Transcription factor SOX8
  • MGC24837|SOX 8|Sox8|SOX8_HUMAN|SRY (sex determining region Y) box 8|SRY box 8|SRY box containing gene 8|Transcription factor SOX 8|Transcription factor SOX-8|Transcription factor SOX8
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P57073
  • P57073
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together