Anti-PSAP Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: PSAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77795-685
- 77795-686
- Antigen symbol:
- PSAP
- PSAP
- Antigen name:
- Prosaposin protein (311-391AA)
- Prosaposin protein (311-391AA)
- Antigen synonyms:
- A1 activator|Cerebroside sulfate activator|Co-beta-glucosidase|Component C|CSAct|Dispersin|GLBA|Glucosylceramidase activator|Proactivator polypeptide|Proactivator polypeptide precursor|Prosaposin (sphingolipid activator protein 1)|prosaposin (variant Gaucher disease and variant metachromatic leukodystrophy)|Prosaposin|Protein A|Protein C|PSAP|SAP 1|SAP 2|SAP-1|SAP-2|SAP_HUMAN|SAP1|Saposin A|Saposin B|Saposin B Val|Saposin C|Saposin D|Saposin-D|Saposins|Sgp1|Sphingolipid activator protein 1|Sphingolipid activator protein 2|Sulfated glycoprotein 1|Sulfatide/GM1 activator
- A1 activator|Cerebroside sulfate activator|Co-beta-glucosidase|Component C|CSAct|Dispersin|GLBA|Glucosylceramidase activator|Proactivator polypeptide|Proactivator polypeptide precursor|Prosaposin (sphingolipid activator protein 1)|prosaposin (variant Gaucher disease and variant metachromatic leukodystrophy)|Prosaposin|Protein A|Protein C|PSAP|SAP 1|SAP 2|SAP-1|SAP-2|SAP_HUMAN|SAP1|Saposin A|Saposin B|Saposin B Val|Saposin C|Saposin D|Saposin-D|Saposins|Sgp1|Sphingolipid activator protein 1|Sphingolipid activator protein 2|Sulfated glycoprotein 1|Sulfatide/GM1 activator
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07602
- P07602
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together