Anti-SERPINF2 Rabbit Polyclonal Antibody
Catalog # 77799-213
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:A2AP
- Antigen symbol:SERPINF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P08697
- Antigen synonyms:A2AP|A2AP_HUMAN|AAP|Alpha 2 antiplasmin|Alpha 2 antiplasmin pigment epithelium derived factor|Alpha 2 AP|ALPHA 2 PI|Alpha 2 plasmin inhibitor|Alpha 2 plasmin inhibitor deficiency|Alpha-2-antiplasmin|Alpha-2-AP|Alpha-2-PI|Alpha-2-plasmin inhibitor|Antiplasmin deficiency|API|Plasmin inhibitor deficiency|PLI|Serine (or cysteine) peptidase inhibitor|clade F|member 2|isoform CRA_c|Serine (or cysteine) proteinase inhibitor|clade F (alpha 2 antiplasmin pigment epithelium derived factor) member 2|Serine or cysteine peptidase inhibitor clade F member 2|Serpin F2|Serpin peptidase inhibitor clade F|Serpin peptidase inhibitor|SERPINF2
- Storage buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SERPINF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat