Anti-RBP5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: RBP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Specifications
- Catalog No:
- 77796-866
- 77796-865
- Antigen symbol:
- RBP5
- RBP5
- Antigen name:
- Retinol-binding protein 5 protein (1-135AA)
- Retinol-binding protein 5 protein (1-135AA)
- Antigen synonyms:
- Cellular retinol-binding protein III|CRBP III|CRBP-III|CRBP3|CRBPIII|HRBPiso|OTTHUMP00000238163|Putative cellular retinol binding protein CRBP III|RBP5|RET5_HUMAN|Retinol binding protein|Retinol binding protein 5|cellular|Retinol-binding protein 5
- Cellular retinol-binding protein III|CRBP III|CRBP-III|CRBP3|CRBPIII|HRBPiso|OTTHUMP00000238163|Putative cellular retinol binding protein CRBP III|RBP5|RET5_HUMAN|Retinol binding protein|Retinol binding protein 5|cellular|Retinol-binding protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P82980
- P82980
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together