Anti-MTMR3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MTMR3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-783
- 77824-782
- Antigen symbol:
- MTMR3
- MTMR3
- Antigen name:
- Myotubularin-related protein 3 protein (652-899AA)
- Myotubularin-related protein 3 protein (652-899AA)
- Antigen synonyms:
- FYVE domain-containing dual specificity protein phosphatase 1|FYVE-DSP1|MTMR3|MTMR3_HUMAN|Myotubularin-related protein 3|Zinc finger FYVE domain-containing protein 10
- FYVE domain-containing dual specificity protein phosphatase 1|FYVE-DSP1|MTMR3|MTMR3_HUMAN|Myotubularin-related protein 3|Zinc finger FYVE domain-containing protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13615
- Q13615
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together