Anti-TCN2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TCN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77802-156
- 77802-155
- Antigen symbol:
- TCN2
- TCN2
- Antigen name:
- Transcobalamin-2 protein (1-200AA)
- Transcobalamin-2 protein (1-200AA)
- Antigen synonyms:
- D22S676|D22S750|II|Macrocytic anemia|OTTHUMP00000199117|OTTHUMP00000199118|OTTHUMP00000199119|TC|TC II|TC II deficiency|TC-2|TC2|TCII|TCN2|TCN2 deficiency|TCO2_HUMAN|Transcobalamin 2|Transcobalamin 2 deficiency|Transcobalamin II|macrocytic anemia|Transcobalamin-2|Vitamin B12 binding protein 2
- D22S676|D22S750|II|Macrocytic anemia|OTTHUMP00000199117|OTTHUMP00000199118|OTTHUMP00000199119|TC|TC II|TC II deficiency|TC-2|TC2|TCII|TCN2|TCN2 deficiency|TCO2_HUMAN|Transcobalamin 2|Transcobalamin 2 deficiency|Transcobalamin II|macrocytic anemia|Transcobalamin-2|Vitamin B12 binding protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20062
- P20062
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together