Anti-SLC26A6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC26A6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-308
- 77835-307
- Antigen symbol:
- SLC26A6
- SLC26A6
- Antigen name:
- Solute carrier family 26 member 6 protein (554-668AA)
- Solute carrier family 26 member 6 protein (554-668AA)
- Antigen synonyms:
- Anion exchange transporter|Anion transporter 1|DKFZp586E1422|Pendrin L1|Pendrin like protein 1|Pendrin-L1|Pendrin-like protein 1|S26A6_HUMAN|SLC26A6|Solute carrier family 26 member 6|Sulfate anion transporter
- Anion exchange transporter|Anion transporter 1|DKFZp586E1422|Pendrin L1|Pendrin like protein 1|Pendrin-L1|Pendrin-like protein 1|S26A6_HUMAN|SLC26A6|Solute carrier family 26 member 6|Sulfate anion transporter
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXS9
- Q9BXS9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together