Anti-SPATA19 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPATA19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-098
- 77832-099
- Antigen symbol:
- SPATA19
- SPATA19
- Antigen name:
- Spermatogenesis-associated protein 19, mitochondrial protein (25-167AA)
- Spermatogenesis-associated protein 19, mitochondrial protein (25-167AA)
- Antigen synonyms:
- SPATA19|SPERGEN1|Spermatogenesis-associated protein 19|mitochondrial|Spermatogenic cell-specific gene 1 protein|Spergen-1
- SPATA19|SPERGEN1|Spermatogenesis-associated protein 19|mitochondrial|Spermatogenic cell-specific gene 1 protein|Spergen-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z5L4
- Q7Z5L4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together