Anti-rbcL Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: rbcL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Soybean
Specifications
- Catalog No:
- 77815-822
- 77815-823
- Antigen symbol:
- rbcL
- rbcL
- Antigen name:
- Recombinant Soybean Ribulose bisphosphate carboxylase large chain protein (3-475AA)
- Recombinant Soybean Ribulose bisphosphate carboxylase large chain protein (3-475AA)
- Antigen synonyms:
- Ribulose bisphosphate carboxylase large chain|RuBisCO large subunit|EC 4.1.1.39
- Ribulose bisphosphate carboxylase large chain|RuBisCO large subunit|EC 4.1.1.39
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Soy bean
- Soy bean
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P27066
- P27066
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together