Anti-FUT1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: FUT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-508
- 77830-507
- Antigen symbol:
- FUT1
- FUT1
- Antigen name:
- Synthetic peptide of FUT1
- Synthetic peptide of FUT1
- Antigen synonyms:
- 2)FT 1|2-alpha-L-fucosyltransferase|Alpha (1 2) fucosyltransferase|Alpha(1 2)FT 1|Alpha(1|Blood group H alpha 2-fucosyltransferase|fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase)|Fucosyltransferase 1|FUT1|FUT1_HUMAN|Galactoside 2 alpha L fucosyltransferase|Galactoside 2-alpha-L-fucosyltransferase 1|GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1|H|HH|HSC|Para Bombay phenotype
- 2)FT 1|2-alpha-L-fucosyltransferase|Alpha (1 2) fucosyltransferase|Alpha(1 2)FT 1|Alpha(1|Blood group H alpha 2-fucosyltransferase|fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase)|Fucosyltransferase 1|FUT1|FUT1_HUMAN|Galactoside 2 alpha L fucosyltransferase|Galactoside 2-alpha-L-fucosyltransferase 1|GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1|H|HH|HSC|Para Bombay phenotype
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19526
- P19526
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together