Anti-IL20RA Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: IL20RA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-742
- 77842-741
- Antigen symbol:
- IL20RA
- IL20RA
- Antigen name:
- Interleukin-20 receptor subunit alpha protein (30-250AA)
- Interleukin-20 receptor subunit alpha protein (30-250AA)
- Antigen synonyms:
- class II cytokine receptor ZCYTOR7|CRF2 8|CRF2-8|Cytokine receptor class II member 8|Cytokine receptor class-II member 8|Cytokine receptor family 2 member 8|I20RA_HUMAN|IL 20R alpha|IL 20R1|IL-20 receptor subunit alpha|IL-20R-alpha|IL-20R1|IL-20RA|IL20 Receptor alpha|IL20RA|Interleukin 20 receptor alpha chain|Interleukin 20 receptor, alpha|interleukin-20 receptor I|Interleukin-20 receptor subunit alpha|ZcytoR7
- class II cytokine receptor ZCYTOR7|CRF2 8|CRF2-8|Cytokine receptor class II member 8|Cytokine receptor class-II member 8|Cytokine receptor family 2 member 8|I20RA_HUMAN|IL 20R alpha|IL 20R1|IL-20 receptor subunit alpha|IL-20R-alpha|IL-20R1|IL-20RA|IL20 Receptor alpha|IL20RA|Interleukin 20 receptor alpha chain|Interleukin 20 receptor, alpha|interleukin-20 receptor I|Interleukin-20 receptor subunit alpha|ZcytoR7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHF4
- Q9UHF4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together