Anti-RANBP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RANBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-629
- 77796-628
- Antigen symbol:
- RANBP2
- RANBP2
- Antigen name:
- E3 SUMO-protein ligase RanBP2 protein (2601-2802AA)
- E3 SUMO-protein ligase RanBP2 protein (2601-2802AA)
- Antigen synonyms:
- 358 kDa nucleoporin|ANE1|E3 SUMO-protein ligase RanBP2|IIAE3|Nuclear pore complex protein Nup358|Nucleoporin 358|Nucleoporin Nup358|NUP358|p270|RAN binding protein 2|Ran-binding protein 2|RBP2_HUMAN|Transformation related protein 2|TRP 1|TRP 2|TRP1|TRP2
- 358 kDa nucleoporin|ANE1|E3 SUMO-protein ligase RanBP2|IIAE3|Nuclear pore complex protein Nup358|Nucleoporin 358|Nucleoporin Nup358|NUP358|p270|RAN binding protein 2|Ran-binding protein 2|RBP2_HUMAN|Transformation related protein 2|TRP 1|TRP 2|TRP1|TRP2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49792
- P49792
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together