Anti-ACAP1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77824-923
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1 protein (1-266AA)
- Antigen symbol:ACAP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15027
- Antigen synonyms:ACAP 1|ACAP1|ACAP1_HUMAN|Arf GAP with coiled coil ANK repeat and PH domains 1|Arf GAP with coiled coil|ankyrin repeat|and pleckstrin homology domains 1|Arf-GAP with coiled-coil|ANK repeat and PH domain-containing protein 1|ArfGAP with coiled-coil|ankyrin repeat and PH domains 1|CENT B1|Centaurin beta 1|Centaurin-beta-1|CENTB 1|Cnt b1|Cnt-b1|Cntb1|KIAA0050
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ACAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse