Anti-SKIL Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SKIL
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-663
- 77799-662
- Antigen symbol:
- SKIL
- SKIL
- Antigen name:
- Ski-like protein (359-579AA)
- Ski-like protein (359-579AA)
- Antigen synonyms:
- OTTHUMP00000213557|OTTHUMP00000213591|OTTHUMP00000213592|OTTHUMP00000213595|SKI like|SKI like oncogene|Ski like protein|SKI like proto oncogene|Ski related oncogene|Ski related oncogene snoN|Ski related protein|Ski-like protein|Ski-related oncogene|Ski-related protein|SKIL|SKIL_HUMAN|SNO|SnoA|SnoI|SnoN
- OTTHUMP00000213557|OTTHUMP00000213591|OTTHUMP00000213592|OTTHUMP00000213595|SKI like|SKI like oncogene|Ski like protein|SKI like proto oncogene|Ski related oncogene|Ski related oncogene snoN|Ski related protein|Ski-like protein|Ski-related oncogene|Ski-related protein|SKIL|SKIL_HUMAN|SNO|SnoA|SnoI|SnoN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12757
- P12757
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together