Anti-PDSS2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: PDSS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-445
- 77832-444
- Antigen symbol:
- PDSS2
- PDSS2
- Antigen name:
- Decaprenyl-diphosphate synthase subunit 2 protein (292-387AA)
- Decaprenyl-diphosphate synthase subunit 2 protein (292-387AA)
- Antigen synonyms:
- All-trans-decaprenyl-diphosphate synthase subunit 2| bA59I9.3| C6orf210| Candidate tumor suppressor protein| chromosome 6 open reading frame 210| Decaprenyl pyrophosphate synthase subunit 2| decaprenyl pyrophosphate synthetase subunit 2 | Decaprenyl-diphosphate synthase subunit 2| DLP1| DLP1_HUMAN| hDLP1| Pdss2| prenyl (decaprenyl) diphosphate synthase| subunit 2| subunit 2 of decaprenyl diphosphate synthase
- All-trans-decaprenyl-diphosphate synthase subunit 2| bA59I9.3| C6orf210| Candidate tumor suppressor protein| chromosome 6 open reading frame 210| Decaprenyl pyrophosphate synthase subunit 2| decaprenyl pyrophosphate synthetase subunit 2 | Decaprenyl-diphosphate synthase subunit 2| DLP1| DLP1_HUMAN| hDLP1| Pdss2| prenyl (decaprenyl) diphosphate synthase| subunit 2| subunit 2 of decaprenyl diphosphate synthase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YH6
- Q86YH6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together