Anti-CEP126 Rabbit Recombinant Polyclonal Antibody
Catalog # 77837-712
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Centrosomal protein of 126 kDa protein (559-670AA)
- Antigen symbol:CEP126
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9P2H0
- Antigen synonyms:CEP126|KIAA1377|Centrosomal protein of 126 kDa
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CEP126
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human