Anti-RBMS1 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: RBMS1
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77796-830
- 77796-829
- Antigen symbol:
- RBMS1
- RBMS1
- Antigen name:
- RNA-binding motif, single-stranded-interacting protein 1 protein (206-277AA)
- RNA-binding motif, single-stranded-interacting protein 1 protein (206-277AA)
- Antigen synonyms:
- c myc gene single strand binding protein 2|C2orf12|Cervical cancer oncogene 4|HCC 4|MSSP 2|MSSP 3|MSSP|MSSP-1|MSSP-2|MSSP-3|MSSP1|RBMS1|RBMS1_HUMAN|RNA binding motif|single stranded interacting protein 1|RNA-binding motif|SCR2|Single stranded DNA binding protein MSSP 1|Single-stranded DNA-binding protein MSSP-1|single-stranded-interacting protein 1|suppressor of cdc 2 (cdc13) with RNA binding motif 2|Suppressor of CDC2 with RNA binding motif 2|Suppressor of CDC2 with RNA-binding motif 2|YC1
- c myc gene single strand binding protein 2|C2orf12|Cervical cancer oncogene 4|HCC 4|MSSP 2|MSSP 3|MSSP|MSSP-1|MSSP-2|MSSP-3|MSSP1|RBMS1|RBMS1_HUMAN|RNA binding motif|single stranded interacting protein 1|RNA-binding motif|SCR2|Single stranded DNA binding protein MSSP 1|Single-stranded DNA-binding protein MSSP-1|single-stranded-interacting protein 1|suppressor of cdc 2 (cdc13) with RNA binding motif 2|Suppressor of CDC2 with RNA binding motif 2|Suppressor of CDC2 with RNA-binding motif 2|YC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29558
- P29558
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together