Anti-PPP3R2 Rabbit Polyclonal Antibody
Catalog # 77795-138
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Protein phosphatase 3 regulatory subunit B beta isoform
- Antigen symbol:PPP3R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96LZ3
- Antigen synonyms:Calcineurin B like protein|Calcineurin B type II (19kDa)|Calcineurin B type II|Calcineurin B-like protein|Calcineurin BII|Calcineurin subunit B isoform 2|Calcineurin subunit B type 2|CANB2_HUMAN|CBLP|CBLP like|CNBII|PPP3R1-like|Ppp3r2|PPP3RL|Protein phosphatase 2B regulatory subunit 2|Protein phosphatase 3 (formerly 2B) regulatory subunit B (19kD) beta isoform (calcineurin B type II)|Protein phosphatase 3 (formerly 2B) regulatory subunit B beta isoform|Protein phosphatase 3 (formerly 2B)|regulatory subunit B|19kDa|beta isoform (calcineurin B|type II)|Protein phosphatase 3 regulatory subunit B (19kD) beta isoform (calcineurin B type II)|Protein phosphatase 3 regulatory subunit B (calcineurin B) like|Protein phosphatase 3 regulatory subunit B beta|Protein phosphatase 3 regulatory subunit B beta isoform
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP3R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat