Anti-SHOX2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SHOX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-527
- 77799-528
- Antigen symbol:
- SHOX2
- SHOX2
- Antigen name:
- Short stature homeobox protein 2 protein (72-331AA)
- Short stature homeobox protein 2 protein (72-331AA)
- Antigen synonyms:
- Homeobox protein Og12X|OG 12|OG 12X|OG12|OG12X|OGI 2X|OGI2X|Paired related homeobox protein SHOT|Paired-related homeobox protein SHOT|Short stature homeobox 2|Short stature homeobox homolog|Short stature homeobox protein 2|SHOT|SHOX 2|SHOX homologous gene on chromosome 3|SHOX2_HUMAN
- Homeobox protein Og12X|OG 12|OG 12X|OG12|OG12X|OGI 2X|OGI2X|Paired related homeobox protein SHOT|Paired-related homeobox protein SHOT|Short stature homeobox 2|Short stature homeobox homolog|Short stature homeobox protein 2|SHOT|SHOX 2|SHOX homologous gene on chromosome 3|SHOX2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60902
- O60902
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together