Anti-SSSCA1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77801-129
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Sjoegren syndrome/scleroderma autoantigen 1 protein (2-199AA)
- Antigen symbol:SSSCA1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O60232
- Antigen synonyms:1500016H19Rik|Autoantigen p27|C184L|centromeric autoantigen (27kD)|OTTHUMP00000232769|OTTHUMP00000233069|OTTHUMP00000233071|p27|Sjoegren syndrome/scleroderma autoantigen 1|Sjogren syndrome/scleroderma autoantigen 1|Sjogren's syndrome/scleroderma autoantigen 1 homolog|SSA27_HUMAN|SSSCA1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: SSSCA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human