Anti-EME1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EME1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-938
- 77833-937
- Antigen symbol:
- EME1
- EME1
- Antigen name:
- Crossover junction endonuclease EME1 protein (1-583AA)
- Crossover junction endonuclease EME1 protein (1-583AA)
- Antigen synonyms:
- 6820428D13| Crossover junction endonuclease EME1| EC 3.1.22.-| EME1| Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)| S. pombe| homolog of| 1| EME1_HUMAN| essential meiotic endonuclease 1 homolog 1 (S pombe)| Essential Meiotic Endonuclease 1 Homolog 1| essential meiotic endonuclease 1 homolog 2| Essential meiotic endonuclease 1| FLJ31364| hMMS4| homolog of yeast EME1 endonuclease| MGC106543| MMS4 | MMS4 homolog| MMS4L
- 6820428D13| Crossover junction endonuclease EME1| EC 3.1.22.-| EME1| Eme1 essential meiotic endonuclease 1 homolog 1 (S. pombe)| S. pombe| homolog of| 1| EME1_HUMAN| essential meiotic endonuclease 1 homolog 1 (S pombe)| Essential Meiotic Endonuclease 1 Homolog 1| essential meiotic endonuclease 1 homolog 2| Essential meiotic endonuclease 1| FLJ31364| hMMS4| homolog of yeast EME1 endonuclease| MGC106543| MMS4 | MMS4 homolog| MMS4L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96AY2
- Q96AY2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together