Anti-TRIP13 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: TRIP13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77825-003
- 77825-002
- Antigen symbol:
- TRIP13
- TRIP13
- Antigen name:
- Pachytene checkpoint protein 2 homolog protein (181-390AA)
- Pachytene checkpoint protein 2 homolog protein (181-390AA)
- Antigen synonyms:
- 16E1-BP| 16E1BP| Homo sapiens HPV16 E1 protein binding protein mRNA complete cds| HPV16 E1 protein binding protein| HPV16 E1 protein-binding protein| Human papillomavirus type 16 E1 protein binding protein| Human papillomavirus type 16 E1 protein-binding protein| Pachytene checkpoint protein 2 homolog| PCH2| Thyroid hormone receptor interactor 13| Thyroid receptor interacting protein 13| Thyroid receptor-interacting protein 13| TR-interacting protein 13| TRIP-13| Trip13| TRP13_HUMAN
- 16E1-BP| 16E1BP| Homo sapiens HPV16 E1 protein binding protein mRNA complete cds| HPV16 E1 protein binding protein| HPV16 E1 protein-binding protein| Human papillomavirus type 16 E1 protein binding protein| Human papillomavirus type 16 E1 protein-binding protein| Pachytene checkpoint protein 2 homolog| PCH2| Thyroid hormone receptor interactor 13| Thyroid receptor interacting protein 13| Thyroid receptor-interacting protein 13| TR-interacting protein 13| TRIP-13| Trip13| TRP13_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15645
- Q15645
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together