Anti-PHF7 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: PHF7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-024
- 77794-025
- Antigen symbol:
- PHF7
- PHF7
- Antigen name:
- PHD finger protein 7 protein (277-368AA)
- PHD finger protein 7 protein (277-368AA)
- Antigen synonyms:
- DKFZp434L1850|HSPC045|HSPC226|MGC26088|NYD SP6|OTTHUMP00000212988|OTTHUMP00000212989|OTTHUMP00000212990|OTTHUMP00000212993|PHD finger protein 7|Phf7|PHF7_HUMAN|Testis development protein NYD SP6|Testis development protein NYD-SP6
- DKFZp434L1850|HSPC045|HSPC226|MGC26088|NYD SP6|OTTHUMP00000212988|OTTHUMP00000212989|OTTHUMP00000212990|OTTHUMP00000212993|PHD finger protein 7|Phf7|PHF7_HUMAN|Testis development protein NYD SP6|Testis development protein NYD-SP6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BWX1
- Q9BWX1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together