Anti-ARHGEF6 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: ARHGEF6
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-617
- 77823-616
- Antigen symbol:
- ARHGEF6
- ARHGEF6
- Antigen name:
- Rho guanine nucleotide exchange factor 6 protein (108-239AA)
- Rho guanine nucleotide exchange factor 6 protein (108-239AA)
- Antigen synonyms:
- Alpha Pix|Alpha-Pix|ARHG6_HUMAN|ARHGEF6|COOL 2|COOL-2|COOL2|Mental retardation|X linked 46|MRX46|PAK interacting exchange factor alpha|PAK-interacting exchange factor alpha|PIXA|Rac/Cdc42 guanine exchange factor (GEF) 6|Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6|Rac/Cdc42 guanine nucleotide exchange factor 6|Rho guanine nucleotide exchange factor 6
- Alpha Pix|Alpha-Pix|ARHG6_HUMAN|ARHGEF6|COOL 2|COOL-2|COOL2|Mental retardation|X linked 46|MRX46|PAK interacting exchange factor alpha|PAK-interacting exchange factor alpha|PIXA|Rac/Cdc42 guanine exchange factor (GEF) 6|Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6|Rac/Cdc42 guanine nucleotide exchange factor 6|Rho guanine nucleotide exchange factor 6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15052
- Q15052
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together