Anti-RAD1 Rabbit Recombinant Polyclonal Antibody
77796-546
New Product
- Antigen name:Cell cycle checkpoint protein RAD1 protein (1-282AA)
- Antigen symbol:RAD1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O60671
- Antigen synonyms:Cell cycle checkpoint protein Hrad1| Cell cycle checkpoint protein Rad 1 A / B| Cell cycle checkpoint protein RAD1| Checkpoint control protein HRAD1| Checkpoint control protein RAD1| DNA repair exonuclease| DNA repair exonuclease rad1| DNA repair exonuclease rad1 homolog| DNA repair exonuclease REC1| DNA repair protein RAD1| EC 3.1.11.2| Exonuclease homolog RAD1| GTP-binding protein RAD| hRAD 1| hRAD1| MGC77779| RAD 1| RAD| RAD1| RAD1 homolog (S. pombe)| RAD1 homolog| Rad1 like DNA damage checkpoint| Rad1 like DNA damage checkpoint protein| S. pombe| homolog of| Rad1-like DNA damage checkpoint protein| RAD1_HUMAN| Ras associated with diabetes| REC 1| REC1| RRAD
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: RAD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human