Anti-UBXN11 Rabbit Polyclonal Antibody
Catalog # 77803-915
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:COA 1
- Antigen symbol:UBXN11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q5T124
- Antigen synonyms:COA 1|colorectal tumor associated antigen 1|Colorectal tumor associated antigen COA 1|Colorectal tumor-associated antigen COA-1|PP2243|SOC|SOCI|Socius|UBX domain containing 5|UBX domain containing protein 11|UBX domain containing protein 5|UBX domain-containing protein 11|UBX domain-containing protein 5|UBX11_HUMAN|UBXD 5|UBXN11
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: UBXN11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat