Anti-CHMP1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHMP1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-831
- 77840-832
- Antigen symbol:
- CHMP1A
- CHMP1A
- Antigen name:
- Charged multivesicular body protein 1a protein (1-196AA)
- Charged multivesicular body protein 1a protein (1-196AA)
- Antigen synonyms:
- Charged multivesicular body protein 1a|CHM1A_HUMAN|CHMP1a|Chromatin modifying protein 1a|Chromatin-modifying protein 1a|hVps46 1|hVps46-1|KIAA0047|Metalloprotease 1|PCOLN3|Procollagen (type III) N endopeptidase|PRSM1|Vacuolar protein sorting 46 1|Vacuolar protein sorting-associated protein 46-1|Vps46 1|Vps46-1
- Charged multivesicular body protein 1a|CHM1A_HUMAN|CHMP1a|Chromatin modifying protein 1a|Chromatin-modifying protein 1a|hVps46 1|hVps46-1|KIAA0047|Metalloprotease 1|PCOLN3|Procollagen (type III) N endopeptidase|PRSM1|Vacuolar protein sorting 46 1|Vacuolar protein sorting-associated protein 46-1|Vps46 1|Vps46-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HD42
- Q9HD42
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together