Anti-WAPL Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: WAPL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-297
- 77832-296
- Antigen symbol:
- WAPL
- WAPL
- Antigen name:
- Wings apart-like protein homolog protein (1037-1190AA)
- Wings apart-like protein homolog protein (1037-1190AA)
- Antigen synonyms:
- FOE|Friend of EBNA2 (Epstein Barr virus nuclear protein 2)|Friend of EBNA2 protein|KIAA0261|OTTHUMP00000020004|WAPAL|WAPL|WAPL_HUMAN|Wings apart like homolog|Wings apart like homolog (Drosophila)|Wings apart-like protein homolog
- FOE|Friend of EBNA2 (Epstein Barr virus nuclear protein 2)|Friend of EBNA2 protein|KIAA0261|OTTHUMP00000020004|WAPAL|WAPL|WAPL_HUMAN|Wings apart like homolog|Wings apart like homolog (Drosophila)|Wings apart-like protein homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z5K2
- Q7Z5K2
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together