Anti-EEF1G Rabbit Polyclonal Antibody
Catalog # 77820-632
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from internal of EEF1G
- Antigen symbol:EEF1G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P26641
- Antigen synonyms:2610301D06Rik|AA407312|eEF 1B gamma|EEF 1G|eEF-1B gamma|EEF1G|EF 1 gamma|EF 1G|EF-1-gamma|EF1 gamma|EF1G|EF1G_HUMAN|Elongation factor 1 gamma|Elongation factor 1-gamma|Eukaryotic translation elongation factor 1 gamma|GIG 35|GIG35|MGC103354|MGC114210|MGC144723|MGC144724|MGC94929|Pancreatic tumor related protein|PRO1608|Translation elongation factor eEF 1 gamma chain
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: EEF1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse