Anti-WIPI1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: WIPI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77827-587
- 77827-588
- Antigen symbol:
- WIPI1
- WIPI1
- Antigen name:
- WD repeat domain phosphoinositide-interacting protein 1 protein (187-446AA)
- WD repeat domain phosphoinositide-interacting protein 1 protein (187-446AA)
- Antigen synonyms:
- ATG 18|ATG18|Atg18 protein homolog|ATG18A|FLJ10055|WD repeat domain phosphoinositide interacting 1|WD repeat domain phosphoinositide interacting protein 1|WD repeat domain phosphoinositide-interacting protein 1|WD40 repeat protein interacting with phosphoinositides of 49 kDa|WD40 repeat protein interacting with phosphoInositides of 49kDa|WIPI 1 alpha|WIPI 1|WIPI 49|WIPI 49 kDa|WIPI-1|wipi1|WIPI1_HUMAN|WIPI49
- ATG 18|ATG18|Atg18 protein homolog|ATG18A|FLJ10055|WD repeat domain phosphoinositide interacting 1|WD repeat domain phosphoinositide interacting protein 1|WD repeat domain phosphoinositide-interacting protein 1|WD40 repeat protein interacting with phosphoinositides of 49 kDa|WD40 repeat protein interacting with phosphoInositides of 49kDa|WIPI 1 alpha|WIPI 1|WIPI 49|WIPI 49 kDa|WIPI-1|wipi1|WIPI1_HUMAN|WIPI49
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5MNZ9
- Q5MNZ9
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together