Order Entry
Canada
ContactUsLinkComponent
Anti-CLPP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-CLPP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CLPP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77823-292
  • 77823-291
  • Antigen symbol:
  • CLPP
  • CLPP
  • Antigen name:
  • ATP-dependent Clp protease proteolytic subunit, mitochondrial protein (223-277AA)
  • ATP-dependent Clp protease proteolytic subunit, mitochondrial protein (223-277AA)
  • Antigen synonyms:
  • ATP dependent protease ClpAP (E coli) proteolytic subunit|ATP dependent protease ClpAP proteolytic subunit|ATP dependent protease ClpAP proteolytic subunit human|ATP dependent proteolytic subunit homolog (E coli)|ATP dependent proteolytic subunit homolog|Caseinolytic peptidase ATP dependent proteolytic subunit homolog|Caseinolytic protease ATP dependent proteolytic subunit E coli|clpP|ClpP caseinolytic peptidase ATP dependent proteolytic subunit|ClpP caseinolytic peptidase ATP dependent proteolytic subunit homolog|CLPP_HUMAN|Endopeptidase Clp|mitochondrial|Putative ATP dependent Clp protease proteolytic subunit mitochondrial|Putative ATP dependent Clp protease proteolytic subunit|Putative ATP-dependent Clp protease proteolytic subunit
  • ATP dependent protease ClpAP (E coli) proteolytic subunit|ATP dependent protease ClpAP proteolytic subunit|ATP dependent protease ClpAP proteolytic subunit human|ATP dependent proteolytic subunit homolog (E coli)|ATP dependent proteolytic subunit homolog|Caseinolytic peptidase ATP dependent proteolytic subunit homolog|Caseinolytic protease ATP dependent proteolytic subunit E coli|clpP|ClpP caseinolytic peptidase ATP dependent proteolytic subunit|ClpP caseinolytic peptidase ATP dependent proteolytic subunit homolog|CLPP_HUMAN|Endopeptidase Clp|mitochondrial|Putative ATP dependent Clp protease proteolytic subunit mitochondrial|Putative ATP dependent Clp protease proteolytic subunit|Putative ATP-dependent Clp protease proteolytic subunit
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q16740
  • Q16740
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together