Anti-TRPC5 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TRPC5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-144
- 77818-143
- Antigen symbol:
- TRPC5
- TRPC5
- Antigen name:
- Short transient receptor potential channel 5 protein (764-973AA)
- Short transient receptor potential channel 5 protein (764-973AA)
- Antigen synonyms:
- Htrp 5|Htrp-5|Htrp5|Short transient receptor potential channel 5|Transient receptor potential cation channel subfamily C member 5|Transient receptor potential channel 5|Transient receptor protein 5|TRP 5|TRP-5|TRP5|TRPC 5|TrpC5|TRPC5_HUMAN
- Htrp 5|Htrp-5|Htrp5|Short transient receptor potential channel 5|Transient receptor potential cation channel subfamily C member 5|Transient receptor potential channel 5|Transient receptor protein 5|TRP 5|TRP-5|TRP5|TRPC 5|TrpC5|TRPC5_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UL62
- Q9UL62
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together