Anti-RTCA Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RTCA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-185
- 77819-186
- Antigen symbol:
- RTCA
- RTCA
- Antigen name:
- RNA 3\'-terminal phosphate cyclase protein (167-379AA)
- RNA 3\'-terminal phosphate cyclase protein (167-379AA)
- Antigen synonyms:
- RTCA|RPC|RPC1|RTC1|RTCD1|RNA 3'-terminal phosphate cyclase|RNA cyclase|RNA-3'-phosphate cyclase|EC 6.5.1.4|RNA terminal phosphate cyclase domain-containing protein 1|RTC domain-containing protein 1
- RTCA|RPC|RPC1|RTC1|RTCD1|RNA 3'-terminal phosphate cyclase|RNA cyclase|RNA-3'-phosphate cyclase|EC 6.5.1.4|RNA terminal phosphate cyclase domain-containing protein 1|RTC domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00442
- O00442
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together