Anti-HPCAL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: HPCAL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77801-077
- 77801-076
- Antigen symbol:
- HPCAL1
- HPCAL1
- Antigen name:
- Hippocalcin-like protein 1 protein (2-193AA)
- Hippocalcin-like protein 1 protein (2-193AA)
- Antigen synonyms:
- BDR1|Calcium binding protein BDR1|Calcium-binding protein BDR-1|hippocalcin like 1|Hippocalcin-like protein 1|HLP2|HPCAL1|HPCAL1 protein|HPCL1_HUMAN|REM-1|VILIP-3|Visinin like protein 3|Visinin-like protein 3
- BDR1|Calcium binding protein BDR1|Calcium-binding protein BDR-1|hippocalcin like 1|Hippocalcin-like protein 1|HLP2|HPCAL1|HPCAL1 protein|HPCL1_HUMAN|REM-1|VILIP-3|Visinin like protein 3|Visinin-like protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P37235
- P37235
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together