Anti-SRSF10 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SRSF10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-356
- 77819-355
- Antigen symbol:
- SRSF10
- SRSF10
- Antigen name:
- Serine/arginine-rich splicing factor 10 protein (1-183AA)
- Serine/arginine-rich splicing factor 10 protein (1-183AA)
- Antigen synonyms:
- 40 kDa SR repressor protein|40 kDa SR-repressor protein|Anti TLS associated protein with SR repeats|arginine/serine-rich 13A|FUS interacting protein serine arginine rich 1|FUS interacting protein serine arginine rich 2|FUS interacting serine arginine rich protein 1|FUS-interacting serine-arginine-rich protein 1|FUSIP1|FUSIP2|NSSR|OTTHUMP00000015774|Serine arginine repressor protein 40 kDa|Serine/arginine-rich splicing factor 10|SFRS13|SFRS13A|Splicing factor|Splicing factor arginine serine rich 13|Splicing factor SRp38|SRp38|SRp40|SRrp40|SRS10_HUMAN|TASR|TASR1|TASR2|TLS associated protein TASR1|TLS associated protein TASR2|TLS associated protein with Ser Arg repeats|TLS associated protein with SR repeats|TLS associated serine arginine protein 1|TLS associated serine arginine protein 2|TLS associated serine arginine protein|TLS associated SR protein|TLS-associated protein with Ser-Arg repeats|TLS-associated protein with SR repeats|TLS-associated serine-arginine protein|TLS-associated SR protein
- 40 kDa SR repressor protein|40 kDa SR-repressor protein|Anti TLS associated protein with SR repeats|arginine/serine-rich 13A|FUS interacting protein serine arginine rich 1|FUS interacting protein serine arginine rich 2|FUS interacting serine arginine rich protein 1|FUS-interacting serine-arginine-rich protein 1|FUSIP1|FUSIP2|NSSR|OTTHUMP00000015774|Serine arginine repressor protein 40 kDa|Serine/arginine-rich splicing factor 10|SFRS13|SFRS13A|Splicing factor|Splicing factor arginine serine rich 13|Splicing factor SRp38|SRp38|SRp40|SRrp40|SRS10_HUMAN|TASR|TASR1|TASR2|TLS associated protein TASR1|TLS associated protein TASR2|TLS associated protein with Ser Arg repeats|TLS associated protein with SR repeats|TLS associated serine arginine protein 1|TLS associated serine arginine protein 2|TLS associated serine arginine protein|TLS associated SR protein|TLS-associated protein with Ser-Arg repeats|TLS-associated protein with SR repeats|TLS-associated serine-arginine protein|TLS-associated SR protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75494
- O75494
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together