Anti-PTPRR Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PTPRR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-437
- 77825-438
- Antigen symbol:
- PTPRR
- PTPRR
- Antigen name:
- Receptor-type tyrosine-protein phosphatase R protein (1-412AA)
- Receptor-type tyrosine-protein phosphatase R protein (1-412AA)
- Antigen synonyms:
- Ch-1PTPase|Ch1 PTPase|DKFZp781C1038|EC PTP|ECPTP|FLJ34328|MGC131968|MGC148170|NC PTPCOM1|NC-PTPCOM1|Protein tyrosine phosphatase Cr1PTPase precursor|Protein tyrosine phosphatase NC PTPCOM1|Protein tyrosine phosphatase PCPTP1|Protein tyrosine phosphatase receptor type R|Protein-tyrosine phosphatase PCPTP1|PTP SL|PTPBR7|PTPRQ|PTPRR|PTPRR_HUMAN|PTPSL|R-PTP-R|Receptor type tyrosine protein phosphatase R|Receptor-type tyrosine-protein phosphatase R
- Ch-1PTPase|Ch1 PTPase|DKFZp781C1038|EC PTP|ECPTP|FLJ34328|MGC131968|MGC148170|NC PTPCOM1|NC-PTPCOM1|Protein tyrosine phosphatase Cr1PTPase precursor|Protein tyrosine phosphatase NC PTPCOM1|Protein tyrosine phosphatase PCPTP1|Protein tyrosine phosphatase receptor type R|Protein-tyrosine phosphatase PCPTP1|PTP SL|PTPBR7|PTPRQ|PTPRR|PTPRR_HUMAN|PTPSL|R-PTP-R|Receptor type tyrosine protein phosphatase R|Receptor-type tyrosine-protein phosphatase R
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15256
- Q15256
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together